4-Fluoro-2-methylpyridine 95

Supplier: Matrix Scientific

073948-1G 073948-5G
102533-850EA 123.08 USD
102533-850 102533-852
4-Fluoro-2-methylpyridine 95
4-Fluoro-2-methylpyridine

Hazard: Irritant

Formula: C₆H₆FN
MW: 111.12 g/mol
MDL Number: MFCD04114145
CAS Number: 766-16-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR