4-Chloro-5-methylpyrimidin-2-amine ≥95%

Supplier: Matrix Scientific

073828-250MG 073828-1G 073828-5G
102533-212EA 461.54 USD
102533-212 102533-214 102533-216
4-Chloro-5-methylpyrimidin-2-amine ≥95%
4-Chloro-5-methylpyrimidin-2-amine

Hazard: Irritant

Formula: C₅H₆ClN₃
MDL Number: MFCD00725898
CAS Number: 20090-58-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR