1-Benzyl-4-methylpiperazine hydrochloride ≥95%

Supplier: Matrix Scientific

073475-5G 073475-10G 073475-100G
102530-362EA 38.46 USD
102530-362 102530-364 102530-366
1-Benzyl-4-methylpiperazine hydrochloride ≥95%
1-Benzyl-4-methylpiperazine hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₉ClN₂
CAS Number: 374898-00-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR