2-Chloro-4,6-dimethoxy-1,3,5-triazine ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Chloro-4,6-dimethoxy-s-triazine

073458-100G 073458-500G
102530-274EA 76.92 USD
102530-274 102530-276
2-Chloro-4,6-dimethoxy-1,3,5-triazine ≥95%
2-Chloro-4,6-dimethoxy-1,3,5-triazine

Hazard: Irritant

Formula: C₅H₆ClN₃O₂
MW: 175.57 g/mol
Boiling Pt: 99…100 °C (1 mmHg)
Melting Pt: 75…79 °C
MDL Number: MFCD00075607
CAS Number: 3140-73-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR