5-Bromo-2,4-dichloro-6-methylpyrimidine ≥95%

Supplier: Matrix Scientific

073326-1G 073326-5G
102529-576EA 146.15 USD
102529-576 102529-578
5-Bromo-2,4-dichloro-6-methylpyrimidine ≥95%
5-Bromo-2,4-dichloro-6-methylpyrimidine

Hazard: Irritant

Formula: C₅H₃BrCl₂N₂
MW: 241.9000 g/mol
MDL Number: MFCD00504524
CAS Number: 56745-01-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR