2-Amino-4,6-dichloropyridine ≥95%

Supplier: Matrix Scientific

073113-1G 073113-5G 073113-10G
102528-444EA 178.46 USD
102528-444 102528-446 102528-448
2-Amino-4,6-dichloropyridine ≥95%
2-Amino-4,6-dichloropyridine

Hazard: Irritant

Formula: C₅H₄Cl₂N₂
MDL Number: MFCD04037218
CAS Number: 116632-24-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR