2-Amino-5-iodo-4-methoxypyrimidine ≥95%

Supplier: Matrix Scientific

073048-250MG 073048-1G 073048-5G
102528-124EA 498.46 USD
102528-124 102528-126 102528-128
2-Amino-5-iodo-4-methoxypyrimidine ≥95%
2-Amino-5-iodo-4-methoxypyrimidine

Hazard: Irritant

Formula: C₅H₆IN₃O
MW: 251.0251 g/mol
MDL Number: MFCD11520889
CAS Number: 89322-66-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR