1-Cyclopropyl-6,7-difluoro-4-oxo-1,4-dihydroquinoline-3-carboxylic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 1-Cyclopropyl-6,7-difluoro-1,4-dihydro-4-oxoquinoline-3-carboxylic acid

072967-1G 072967-5G
102527-698EA 176.92 USD
102527-698 102527-700
1-Cyclopropyl-6,7-difluoro-4-oxo-1,4-dihydroquinoline-3-carboxylic acid ≥95%
1-Cyclopropyl-6,7-difluoro-4-oxo-1,4-dihydroquinoline-3-carboxylic acid

Hazard: Irritant

Formula: C₁₃H₉F₂NO₃
MDL Number: MFCD01646375
CAS Number: 93107-30-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR