Methyl-3-hydroxypicolinate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 3-Hydroxy-2-pyridinecarboxylate, 3-Hydroxypyridine-2-carboxylic acid methyl ester, 3-Hydroxypicolinic acid methyll ester, Methyl 3-hydroxypicolinate, 3-Hydroxy-2-pyridinecarboxylic acid methyll ester, Methyl 3-Hydroxypicolinate

072912-1G 072912-5G
102527-402EA 60 USD
102527-402 102527-404
Methyl-3-hydroxypicolinate ≥95%
Methyl-3-hydroxypicolinate

Hazard: Irritant

Formula: C₇H₇NO₃
MW: 153.1354 g/mol
MDL Number: MFCD00661311
CAS Number: 62733-99-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR