4-Aminoquinoline ≥95%
Supplier: Matrix Scientific
072898-1G
072898-5G
072898-10G
102527-332EA
127.69
USD
102527-332
102527-334
102527-336
4-Aminoquinoline ≥95%
4-Aminoquinoline
Hazard: Irritant
Formula:
C₉H₈N₂ |
MDL Number:
MFCD00463448 CAS Number: 578-68-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...