(4-(4-Fluorobenzyl)morpholin-2-yl)methanamine ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Aminomethy-4-(4-fluorobenzyl)morpholine

072700-1G 072700-5G 072700-10G
102526-254EA 90.77 USD
102526-254 102526-256 102526-258
(4-(4-Fluorobenzyl)morpholin-2-yl)methanamine ≥95%
(4-(4-Fluorobenzyl)morpholin-2-yl)methanamine

Hazard: Irritant

Formula: C₁₂H₁₇FN₂O
MW: 224.28 g/mol
MDL Number: MFCD06658145
CAS Number: 112914-13-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR