Ethyl-5-bromopyrazine-2-carboxylate ≥95%

Supplier: Matrix Scientific

072559-250MG 072559-1G 072559-5G
102525-510EA 640 USD
102525-510 102525-512 102525-514
Ethyl-5-bromopyrazine-2-carboxylate ≥95%
Ethyl 5-bromopyrazine-2-carboxylate

Hazard: Irritant

Formula: C₇H₇BrN₂O₂
MDL Number: MFCD11044905
CAS Number: 36070-83-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR