4-Amino-m-anisic acid ≥95%
Supplier: Matrix Scientific
Synonyms:
4-Amino-3-methoxybenzenecarboxylic acid
072526-5G
072526-25G
102525-318EA
73.85
USD
102525-318
102833-532
4-Amino-m-anisic acid ≥95%
4-Amino-m-anisic acid
Hazard: Irritant
Formula:
C₈H₉NO₃ MW: 167.16 Melting Pt: 186…188 °C |
MDL Number:
MFCD00016539 CAS Number: 2486-69-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...