3-Amino-1-methyl-1H-pyrazole-4-carbonitrile ≥95%
Supplier: Matrix Scientific
Synonyms:
1-Methyl-3-amino-4-cyanopyrazole, 3-amino-1-methyl-1H-pyrazole-4-carbonitrile
072518-250MG
072518-1G
072518-5G
102525-268EA
427.69
USD
102525-268
102525-270
102525-272
3-Amino-1-methyl-1H-pyrazole-4-carbonitrile ≥95%
3-Amino-1-methyl-1H-pyrazole-4-carbonitrile
Hazard: Irritant
Formula:
C₅H₆N₄ |
CAS Number:
21230-50-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...