(R)-(+)-α,α-Diphenylprolinol ≥95%

Supplier: Matrix Scientific

Synonyms: (R)-(+)-alpha,alpha-Diphenylprolinol, (R)-(+)-α,α-Diphenyl-2-pyrrolidinemethanol, (R)-(+)-2-(Diphenylhydroxymethyl)pyrrolidine, α,α-Diphenyl-D-prolinol

072371-1G 072371-5G 072371-10G
102524-490EA 15.38 USD
102524-490 102524-492 102524-494
(R)-(+)-α,α-Diphenylprolinol ≥95%
(R)-(+)-α,α-Diphenylprolinol

Hazard: Irritant

Formula: C₁₇H₁₉NO
MDL Number: MFCD00077754
CAS Number: 22348-32-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR