5-Iodopyrazin-2-amine ≥95%
Supplier: Matrix Scientific
072328-1G
072328-5G
102524-274EA
36.92
USD
102524-274
102524-276
5-Iodopyrazin-2-amine ≥95%
5-Iodopyrazin-2-amine
Hazard: Irritant
Formula:
C4H4IN3 |
MDL Number:
MFCD09909691 CAS Number: 886860-50-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...