2-Aminopyridin-4-ol ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Amino-4-hydroxypyridine

072100-5G
102523-094EA 63.08 USD
102523-094
2-Aminopyridin-4-ol ≥95%
2-Aminopyridin-4-ol

Hazard: Irritant

Formula: C₅H₆N₂O
MW: 110.12 g/mol
MDL Number: MFCD04114161
CAS Number: 33631-05-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR