Benzyl-4-oxo-1-piperidinecarboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: 1-Benzyloxycarbonyl-4-piperidine1-Carbobenzoxy-4-piperidone1-Cbz-4-piperidone4-Oxo-1-piperidinecarboxylic Acid Benzyl Ester1-Cbz-Piperidin-4-one

072084-25G 072084-100G
102523-008EA 38.46 USD
102523-008 102523-010
Benzyl-4-oxo-1-piperidinecarboxylate ≥95%
Benzyl-4-oxo-1-piperidinecarboxylate

Purity: 95+%
CAS: 19099-93-5
MDL Number: MFCD00673144
Molecular Formula: C13H15NO3
Molecular Weight: 233,27
Hazard: Irritant

Formula: C₁₃H₁₅NO₃
MW: 233.27 g/mol
Melting Pt: 38…41 °C
Density: 1.172
MDL Number: MFCD00673144
CAS Number: 19099-93-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR