4-Amino-5-bromo-2-chloropyrimidine ≥95%

Supplier: Matrix Scientific

071945-1G 071945-5G 071945-10G
102522-238EA 27.69 USD
102522-238 102522-240 102522-242
4-Amino-5-bromo-2-chloropyrimidine ≥95%
4-Amino-5-bromo-2-chloropyrimidine

Hazard: Irritant

Formula: C₄H₃BrClN₃
MDL Number: MFCD07782040
CAS Number: 205672-25-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR