a-Tosylbenzyl isocyanide ≥95%

Supplier: Matrix Scientific

Synonyms: (1-Phenyl-1-tosyl)methyl Isocyanide, alpha-(p-Toluenesulfonyl)benzyl Isocyanide, α-Tosylbenzyl Isocyanide, α-(p-Toluenesulphonyl)benzyl Isocyanide, alpha-(p-Toluenesulphonyl)benzyl Isocyanide, alpha-Tosylbenzyl Isocyanide, α-(p-Toluenesulfonyl)benzyl Isocyanide

071797-1G 071797-5G 071797-10G
102521-388EA 284.62 USD
102521-388 102521-390 102521-392
a-Tosylbenzyl isocyanide ≥95%
a-Tosylbenzyl isocyanide

Hazard: Irritant

Formula: C₁₅H₁₃NO₂S
MW: 271.3342 g/mol
MDL Number: MFCD04114776
CAS Number: 36635-66-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR