FGE-XY Digital Force Gauge, SEALS USA INC
Supplier: SEALS USA INC
FGE-10XY
97024-778EA 1498.81 USD
97024-778
FGE-XY Digital Force Gauge, SEALS USA INC
Dynamometers
The Series FGE-XY Digital Force Gauges combine a rugged, ergonomic and compact design with exceptional accuracy to provide a solid force testing instrument
Simple one button adjustment alters units, while pressing the peak button quickly produces the highest recorded value during the test. The 180° reversible display combined with the adjustable force direction and dual labeled keypad enable the FGE-XY to excel in both compression and tension applications in any direction. Models are available from 0.5 lbs to 200 lb ranges providing a large offering for various testing requirements.
Typical applications include wallboard, wire pull, wire crimps, trigger pull, brake pedal, aerospace ceramic, springs, food ripeness, corrugated paper strength, blister pack, medical component, muscle strength, ergonomic testing, adhesives, syringe plunger, needle sharpness and many more.
These instruments are utilized in a variety of applications such as, incoming quality inspection, finished goods testing, R&D or almost any portable or force stand testing requirement.
Certifications: CE Compliant.
Ordering information: Carrying Case Included, AC Adapter/Charger Included, Hook, Chisel, Flat Head, Notched Head, Hanger, Cone Head, Thread Adapter, Extension Rod, USB Cable all Included, 2-Year Warranty, NIST Certification Optional.