WaterPro® RO Systems Accessories, Labconco
Supplier: Labconco
9078700 9067201
26301-702EA 2205.2 USD
26301-702 26302-014
WaterPro® RO Systems Accessories, Labconco
Water Purification Systems Pure and Ultra Pure Water Systems
Prefilter/Carbon Filter Kit required for operation of the WaterPro® RO System.
Ordering information: WaterPro® RO System sold separately.
Delivery information: Kit includes Prefilters (3) and Carbon Filters (3) for initial start-up, and two filter changes. The Reverse Osmosis Membrane is required for operation of WaterPro® RO System.