Anti-IgG Goat Antibody (AzureSpectra 650)

Supplier: Azure Biosystems
AC2169
75794-830EA 147.84 USD
75794-830
Anti-IgG Goat Antibody (AzureSpectra 650)
Antibodies
Type: Secondary
Antigen: IgG
Clonality:
Clone:
Conjugation: AzureSpectra 650
Epitope:
Host: Goat
Isotype:
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR