Calcium Standard for AAS, Certipur®, MilliporeSigma
Supplier: MilliporeSigma
1.19778.0100
1.19778.0500
EM1.19778.0100EA
108
USD
EM1.19778.0100
EM1.19778.0500
Calcium Standard for AAS, Certipur®, MilliporeSigma
Standards
Cationic Standards
Calcium Standards
Ca(NO₃)₂ in HNO₃
Traceable to SRM from NIST.