4-Picoline, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: γ-Picoline4-Methylpyridine

M-Clarity™ quality level = MQ200

8.07049.0010 8.07049.0010
EM8.07049.0010EA 27.81 USD
EM8.07049.0010 CA8.07049.0010
4-Picoline, Sigma-Aldrich®
4-Picoline
Formula: C₆H₇N
MW: 93.13 g/mol
Boiling Pt: 144.9 °C (1013 hPa)
Melting Pt: 3 °C
Density: 0.9547 g/cm³ (20 °C)
Flash Pt: 40 °C
MDL Number: MFCD00006440
CAS Number: 108-89-4
UN: 2313
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR