3-Aminophenol, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning

Synonyms: m-Aminophenol3-Hydroxyaniline

M-Clarity™ quality level = MQ200

8.00420.9050 8.00420.9050
EM8.00420.9050EA 6747 USD
EM8.00420.9050 CA8.00420.9050
3-Aminophenol, Sigma-Aldrich®
3-Aminophenol
Formula: C₆H₇NO
MW: 109.13 g/mol
Boiling Pt: 164 °C (15 hPa)
Melting Pt: 122 °C
Density: 0.99 g/cm3 (25 °C)
Flash Pt: 92 °C
Storage Temperature: Ambient
MDL Number: MFCD00007786
CAS Number: 591-27-5
UN: 2512
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR