2-Nitrobenzaldehyde, Sigma-Aldrich®

Supplier: MilliporeSigma


M-Clarity™ quality level = MQ200

8.22293.0001 8.22293.0001
EM8.22293.0001EA 45.32 USD
CA8.22293.0001 EM8.22293.0001
2-Nitrobenzaldehyde, Sigma-Aldrich®
2-Nitrobenzaldehyde
Formula: C₇H₅NO₃
MW: 151.12 g/mol
Boiling Pt: 285 °C (1013 hPa)
Melting Pt: 41…43 °C
Density: 1.35 g/cm³ (25 °C)
MDL Number: MFCD00007132
CAS Number: 552-89-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR