Acrylic acid, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: Prop-2-enoic acid

M-Clarity™ quality level = MQ200

8.00181.9025 8.00181.9025
EM8.00181.9025EA 808 USD
CA8.00181.9025 EM8.00181.9025
Acrylic acid, Sigma-Aldrich®
Acrylic acid
Formula: C₃H₄O₂
MW: 72.06 g/mol
Boiling Pt: 141.6 °C (1013 hPa)
Melting Pt: 13 °C
Density: 1.06 g/cm³ (10 °C)
MDL Number: MFCD00004367
CAS Number: 79-10-7
UN: 2218
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR