2,3-Dimercapto-1-propanol, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: 2,3-DimercaptopropanolDithioglycerineDimercaprolDimercapto-2,3 propanol-12,3-Dimercaptopropane-1-ol2,3-Dimercaptopropan-1-olBAL

M-Clarity™ quality level = MQ200

8.18757.0002 8.18757.0002
EM8.18757.0002EA 160 USD
EM8.18757.0002 CA8.18757.0002
2,3-Dimercapto-1-propanol, Sigma-Aldrich®
2,3-Dimercapto-1-propanol
Formula: C₃H₈OS₂
MW: 124.22 g/mol
Boiling Pt: 223 °C (1013 hPa)
Melting Pt: 77 °C
Density: 1.2531 g/cm³ (20 °C)
Flash Pt: 112 °C (closed cup)
MDL Number: MFCD00004864
CAS Number: 59-52-9
UN: 2810
ADR: 6.1,III
https://us.vwr.com/stibo/web/std.lang.all/81/18/2358118.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR