1-Octanesulfonic acid sodium salt, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: Sodium 1-octansulfonateSodium octane-1-sulphonate

M-Clarity™ quality level = MQ200

1.12292.0002 1.12292.0002
EM1.12292.0002EA 96.82 USD
CA1.12292.0002 EM1.12292.0002
1-Octanesulfonic acid sodium salt, Sigma-Aldrich®
1-Octanesulphonic acid sodium salt
Formula: C₈H₁₇NaO₃S
MW: 216.28 g/mol
Melting Pt: > 300 °C
MDL Number: MFCD00007544
CAS Number: 5324-84-5
https://us.vwr.com/stibo/web/std.lang.all/09/50/2300950.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR