Density and Mobility in N–Germanium/P–Germanium
Supplier: Klinger
P7213 586852
470220-130KT 15834 USD
470220-130 470220-132
Density and Mobility in N–Germanium/P–Germanium
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
Solid-state physics – conduction phenomena.
  • Includes all major components
  • Includes two material options N and P type crystals
Explore the temperature-dependency of the Hall voltage and the electrical conductivity
using doped germanium samples. The concentrations of the charge carriers and their mobility's are determined under the assumption that, depending on the doping, one of the concentrations n or p can be ignored.
Required but not included: PC with Windows XP/Vista/7/8/10 (x86 or x64).
Base unit for Hall Effect
n-Ge on plug-in board or p-GeMore Product Information