Hall Effect Apparatus
Supplier: Klinger
P7211B 58684
470220-126KT 13020.19 USD
470220-126 470220-128
Hall Effect Apparatus
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
The Hall Effect Apparatus explores solid-state physics and conduction phenomena.
  • All Major Components Included
  • Two Material Options: Silver and Tungsten
  • Apparatus Explores Solid-State Physics and Conduction Phenomena
  • Hall Coefficient Determined by Measuring Hall Voltage UH for Various Currents
  • Negative Value Indicates Electrons Are Transporting Voltage
Users determine the Hall coefficient RH of two electrical conductors by measuring the Hall voltage UH for various currents I as a function of the magnetic field B.
A negative value is obtained forMore Product Information