Back Ferrule, Parker, Restek
Supplier: Restek
21907 21806 21807 21808 21906
10855-394EA 136.93 USD
10855-394 10855-396 10855-482 10855-548 10855-550 10855-602
Back Ferrule, Parker, Restek
Fittings and Connectors
Back ferrule of a ferrule set for providing a seal when compressed between two mating components.
  • Connects to tubing
  • Brass construction for corrosion resistance, ductility at high temperatures, and low magnetic permeability
Pipe and tube fittings are components used for connecting, terminating, controlling flow, and changing the direction of piping in many different industries.
Ordering information: Please note: Nuts and ferrules are not treated unless requested (custom parts). Nuts and ferrules normally are not in contact with sample pathway, and thus do not require coating.