Versa Vials™, Glass, Restek
Supplier: Restek
22699 22700 22701 22702 22703 22704
10854-128PK 30.11 USD
10854-128 10854-130 10854-132 10854-134 10854-136 10854-138
Versa Vials™, Glass, Restek
Vials
Versa Vials™ feature a 12 mm cap size and 9 mm opening that is ideal for sampling and pipette use.
  • Clear or amber color
  • Glass fabrication
  • 12 mm orifice
Elongated neck allows for consistent, reliable autosampler pick-up. Vials have a volume of 2 ml and an outside diameter of 12 mm.