PTFE Ferrules, Restek
Supplier: Restek
21122 21125 21126 21128 21127
10058-604PK 53.7 USD
10058-604 10058-606 10058-608 10058-610 10852-614 10852-616 10852-622
PTFE Ferrules, Restek
Fittings and Connectors
PTFE ferrules are built to be extremely durable, with an inert nature and temperature resistance up to 250°C.
  • 100% PTFE fabrication
  • Completely inert
  • One-piece design requires no back ferrule
  • Upper temperature limit of 250°C