Two-Hole Ferrules for ¹/₈ and ¹/₁₆" Compression-Type Fittings, Restek
Supplier: Restek
20246 24848 24849
RK20246PK 50.94 USD
RK20246 10059-310 10059-312
Two-Hole Ferrules for ¹/₈ and ¹/₁₆" Compression-Type Fittings, Restek
Fittings and Connectors
Two-hole ferrules seal with minimal torque, are reusable, and are preferred for vacuum and high-pressure uses.
  • Use 1/16-inch, two-hole ferrules with the 1/16-inch capillary inlet adaptor fitting kit
  • Use 1/8-inch, two-hole ferrules with the 1/8-inch capillary inlet adaptor fitting kit
  • Stable to 400°C
For use with 1/8" and 1/16" compression-type fittings.