Potassium Permanganate

Supplier: ALDON CORP SE

PP0700-100GR PP0700-500GR PP0700-100G PP0700-500G PP0711-1L PP0711-500ML PP0700-PK/4
470302-236EA 12.7 USD
470302-236 470302-238 470302-212 470302-214 470302-216 470302-220 470302-228
Potassium Permanganate
Chemicals for Science Education

CAS Number: 7722-64-7
Formula Weight: 158.04
Formula: KMnO4
Hazard Info: Oxidizer, Explosive, Irritant
Density (g/mL): 2.7032
Boiling Point (°C): Decomposes
Freezing Point (°C): Decomposes
Solubility: Water
Synonyms: Chameleon Mineral
Shelf Life (months): 36
Storage: Yellow


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR