Papain

Supplier: Thermo Scientific Chemicals

41676-0100 41676-5000
200007-270EA 137 USD
200007-270 200013-888 200020-302 TS41676-0100 TS41676-5000
Papain
Enzymes

CAS No.: 9001-73-4


MDL No.: MFCD00131791
GHS Symbols: GHS08
Appearance: Cream to light brown Powder
Storage Temperature: Refrigerator
Signal word: Danger

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR