Carbamazepine 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 5H-Dibenzo[b,f]azepine-5-carboxamide

AC34436-0050 AC34436-0250 AC34436-0010
200024-672EA 188 USD
200024-672 103513-878 200018-424
Carbamazepine 98%
Carbamazepine
Formula: C₁₅H₁₂N₂O
MW: 236.27 g/mol
Melting Pt: 189…192 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005073
CAS Number: 298-46-4
EINECS: 206-062-7
ADR: 3,II

Order Now

Specification Test Results

Appearance White to off-white Powder or crystals
Infrared spectrum Conforms
Melting point 189 to 193 °C
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR