Guanosine-5'-monophosphate disodium salt 97%
Supplier: Thermo Scientific Chemicals
Synonyms:
Disodium 5'-guanylate
, 5'-Guanosine monophosphate disodium salt
, 5'-GMP 2Na
, 5'-Guanylic acid disodium salt
, GMP disodium salt
, Guanosine-5'-Monophosphate
AC22660-0050
AC22660-0250
AC22660-1000
103517-742EA
652.6
USD
103517-742
103517-740
200012-626
Guanosine-5'-monophosphate disodium salt 97%
Guanosine-5'-monophosphate disodium salt
MW:
407.187 g/mol Melting Pt: >300 °C Storage Temperature: Ambient |
EINECS:
226-914-1 |
Specification Test Results
Appearance | White Crystalline powder |
Infrared spectrum | Conforms |
Loss on drying | ≤25 % (120 °C, 4 h) |
TLC | Homogeneous |
UV spectrophotometry | ≥97.0 % (lambda max. 252 nm, pH 7) |
UV spectrophotometry | molar absorptivity 13700 +/- 4%, pH 7 |
Titration with HClO4 | ≥96.0 % (On dry substance) |
Heavy metals (as Pb) | ≤10 ppm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...