Dowex® 50WX2 (H) 100-200 mesh ion-exchange resin

Supplier: Thermo Scientific Chemicals
Danger

AC20303-5000 AC20303-1000 AC20303-0025
200012-286EA 257 USD
200012-286 200018-984 103467-338
Dowex® 50WX2 (H) 100-200 mesh ion-exchange resin
Dowex® 50WX2 (H)
Storage Temperature: Ambient
MDL Number: MFCD00132722
CAS Number: 12612-37-2

Order Now

Specification Test Results

Appearance Beads
Particle size ≤15 % on 35 mesh (cumulative)
Particle size ≤15 % Through 140 mesh
Wet volume capacity ≥0.6 meq/ml H form
Water retention capacity 74 to 82 % H form

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR