Anti-IgG Donkey Polyclonal Antibody (CF™ 405S)

Supplier: Biotium

20416-500UL 20416-50UL
10156-350EA 320.98 USD
10156-350 10018-320
Anti-IgG Donkey Polyclonal Antibody (CF™ 405S)
Antibodies
This product is prepared by labeling highly cross-adsorbed donkey anti-goat IgG (H+L) with CF™405S. CF™405S is a blue fluorescent dye with an absorption peak wavelength that nearly coincides with the 405 nm blue diode laser line. In addition, the emission peak wavelength of the dye well centers within the blue detection window of BD flow cytometers. As a result, flow cytometry analysis using CF™405S dye results in much brighter signal than that using Alexa Fluor® 405. To minimize cross reactivity, the antibody has been adsorbed against chicken, guinea pig, Syrian hamster, horse, human, mouse, rabbit, and rat serum proteins.

Alexa Fluor® is a registered trademark of Thermo Fisher Scientific.

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: CF405S
Epitope:
Host: Donkey
Isotype:
Reactivity: Goat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR