Anti-RFP Rabbit Polyclonal Antibody (CF488A)

Supplier: Biotium

20421
10018-146EA 391.57 USD
10018-146
Anti-RFP Rabbit Polyclonal Antibody (CF488A)
Antibodies
Polyclonal anti-RFP reacts with red fluorescent protein (RFP from Discosoma spp.) and its variants. To minimize cross-reactivity, the antibody has been adsorbed against human, mouse and rat serum proteins. This conjugate is prepared by labeling polyclonal rabbit anti-RFP IgG (H+L) with CF™488A dye. CF™488A (Ex/Em 490/515 nm) is a superior alternative to the traditional green fluorescent dye FITC (or fluorescein) owing to its exceptional brightness and photostability. CF™488A has brightness and photostability comparable to Alexa Fluor® 488.

Alexa Fluor® is a registered trademark of Thermo Fisher Scientific.

Type: Tag
Antigen: RFP
Clonality: Polyclonal
Clone:
Conjugation: CF488A
Epitope:
Host: Rabbit
Isotype:
Reactivity: Discosoma spp.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR