Anti-FLAG Tag Rabbit Polyclonal Antibody (CF594)

Supplier: Biotium

20434
10001-602EA 330.79 USD
10001-602
Anti-FLAG Tag Rabbit Polyclonal Antibody (CF594)
Antibodies
Polyclonal anti-FLAG (DYKDDDDK) reacts with FLAG tagged fusion proteins. The antibody recognizes the epitope tag fused to either the amino- or carboxy- termini of targeted proteins. This conjugate is prepared by labeling polyclonal rabbit anti-FLAG IgG (H+L) with CF™594 dye. CF™594 (Ex/Em 593/614 nm) is a deep-red dye spectrally similar to Alexa Fluor® 594 and Texas Red® dye but yields much brighter protein conjugates due to its high quantum yield and exceptional water solubility. CF™594 also has excellent photostability ideal for demanding applications, such as confocal microscopy and single molecule imaging.

Alexa Fluor® and Texas Red® are registered trademarks of Thermo Fisher Scientific.

Type: Tag
Antigen: FLAG Tag
Clonality: Polyclonal
Clone:
Conjugation: CF594
Epitope:
Host: Rabbit
Isotype:
Reactivity: FLAG tag
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR