Anti-IgG Goat Antibody (Biotin)

Supplier: Vector Laboratories
BA-1000-1.5
101097-810EA 267.23 USD
101097-810
Anti-IgG Goat Antibody (Biotin)
Antibodies
Affiinity-purified, ultrapure, high affinity antibody. Can be used for tissue and cell staining, ELISAs, and blots. This antibody is included in the VECTASTAIN ABC kits.

  • Biotinylated to ensure the maximum degree of labeling without compromising the specificity or affinity of the antibody. Thoroughly adsorbed against serum and immunoglobulins from potentially interfering species. The recommended dilution for most applications is 1:200.

Type: Secondary
Antigen:
Clonality:
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: IgG (H+L)
Reactivity: Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR