Imizedmobil Catalase beads are the ideal solution for HTP preparation of samples containing hydrogen peroxide or unwanted analytes.
- No interference with downstream analyses
- Used to eliminate or reduce sample background from hydrogen peroxide in assays
- Reactions can be carried out with simultaneous removal of hydrogen peroxide
Immobilized Catalase beads are the ideal solution for HTP preparation of samples containing hydrogen peroxide or unwanted analytes which generate hydrogen peroxide that interferes with downstream analyses.
The immobilized catalase beads can be used to eliminate or reduce sample background generated from hydrogen peroxide in assays that also use hydrogen peroxide for signal detection. Centrifugation to remove the immobilized catalase beads will prevent interference of catalase in assays that use hydrogen peroxide/HRP based mechanisms for detection. If unwanted interfering substances are removed during sample prep by oxidation, the reaction can be carried out to completion with simultaneous removal of hydrogen peroxide.
User Supplied Reagents and Equipments include 96-well polypropylene plate and a centrifuge for 96-well plates
Protocol for best use: mix beads to generate homogeneous 50% suspension, add 10 µl of the 50 % slurry of the immobilized catalase beads to 50 µl sample of interest , incubate the sample at room temperature for 15 minutes, spin down the sample at 1000 x g for a minute at RT to remove the beads, then collect the supernatant and use for downstream analyses.