DermaLife MA Adult Melanocyte Medium, Lifeline Cell Technology
Supplier: Lifeline Cell Technology
LL-0039 LS-1063
102970-832EA 280 USD
102970-832 102970-868
DermaLife MA Adult Melanocyte Medium, Lifeline Cell Technology
Cell Culture Media
DermaLife Ma Melanocyte Medium is a low serum, PMA-free, Cholera toxin-free medium optimized for the culture of Human Adult Epidermal Melanocytes
  • Optimized for the culture of Human Adult Epidermal Melanocytes
  • Contains no phenol red or antimicrobials
DermaLife Ma Melanocyte Adult Medium Complete Kit includes: DermaLife Basal Medium 500 mL and DermaLife Ma Melanocyte Adult LifeFactors® Kit. DermaLife Ma LifeFactors® Kit includes the associated supplements and growth factors called "LifeFactors®" for use with human adult melanocytes and DermaLife basal medium, to allow you to prepare fresh medium.
DermaLife contains no phenol red or antimicrobials.
Ordering information: The DermaLife Ma LifeFactors® Kit includes: L-Glutamine 15 mL, Calcium Chloride, Epinephrine 0.5 mL, StiMel8 5 mL, rh Insulin 0.5 mL, Ascorbic Acid 0.5 mL, TM3 1.0 mL, Gentamicin and Amphotericin B 0.5 mL.
DermaLife Medium Complete Kits include DermaLife Basal Medium and the associated LifeFactors® Kit. DermaLife Basal Medium requires the appropriate LifeFactors®.