Compliance Gold® Occult Blood (iFOB) Tests, Germaine® Laboratories
Supplier: Germaine Laboratories
74034 74014 74030 74015 74033 74008
10024-738BX 651.12 USD
10024-738 10024-740 10024-742 10024-806 10024-808 10024-810
Compliance Gold® Occult Blood (iFOB) Tests, Germaine® Laboratories
Clinical Diagnostic Supplies Clinical Reagents and Kits
Compliance Gold® iFOB (immunological fecal occult blood) is a rapid chromatographic assay for the qualitative detection of occult blood in human stool samples
Compliance Gold® tests are packaged in various sizes to fit every need. Some versions allow for the patient to take the test home and return specimens via mail. Complete return mailers include a collection bottle, collection paper, specimen bag, and easy to follow instructions.
The full compliance verison includes a Compliance Monotoring System, during which a team member will provide a HIPAA compliant program that reminds patients to return collection kits.
These tests are used to determine gostrointestinal disorders such as colitis, colorectal cancer, diverticulitis, and polyps.
Certifications: CLIA waived.