AccuHome® Pregnancy OTC Test, Germaine® Laboratories
Supplier: Germaine Laboratories
89101 89102
10024-398EA 8.09 USD
10024-398 75796-392
AccuHome® Pregnancy OTC Test, Germaine® Laboratories
Clinical Diagnostic Supplies Clinical Reagents and Kits
The AccuHome® Pregnancy OTC Test is an over the counter urine midstream pregnancy test
The test features built-in controls to ensure accuracy, and is sensitive to 25 mIU/mL hCG. It provides results in 3 minutes, with 1 line for negative results and 2 lines for positive results.
Certifications: CLIA waived.