Pregnancy Tests, Germaine® Laboratories
Supplier: Germaine Laboratories
88530 97030 97730
10024-388BX 61.35 USD
10024-388 10024-390 10024-394
Pregnancy Tests, Germaine® Laboratories
Clinical Diagnostic Supplies Clinical Reagents and Kits
These pregnancy tests are designed for the qualitative determination of hCG in urine, to aid in the early detection of pregnancy
All tests feature built-in controls to ensure accuracy, and are sensitive to 20 mIU/mL hCG. Results are available in 3 to 5 minutes, with 1 line for negative results and 2 lines for positive results.
Recommended for professional use.
Certifications: CLIA waived.